Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ddTTP (trisodium) solution | 00-00-0 | 99.9% | 532.11 g·mol⁻¹ | C10H14N2Na3O13P3 | 5 MG
SDP

Catalog No. 5000222672
Encompass_Preferred

ddTTP (trisodium) solution (100 mM) is a concentrated aqueous 2',3'-dideoxyribonucleoside 5'-triphosphate used to terminate DNA chain elongation in sequencing and other polymerase reactions. Supplied for research use only as a ready-to-use liquid, it provides a high-purity nucleotide analogue for enzymatic assays.

  • Concentration: 100 mM aqueous solution.
  • High purity: 99.94%.
  • Intended for research use only.
  • Function: chain termination of DNA polymerase in sequencing.
  • Form: liquid, colorless to light yellow.
  • Molecular weight 532.11 g·mol⁻¹; formula C10H14N2Na3O13P3.
  • Available in small prepared aliquots such as 5 mg.

Catalog No. 50-002-22672 Supplier Medchemexpress LLC Supplier No. HY137694B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ddTTP (trisodium) solution (100 mM) is a concentrated aqueous 2',3'-dideoxyribonucleoside 5'-triphosphate used to terminate DNA chain elongation in sequencing and other polymerase reactions. Supplied for research use only as a ready-to-use liquid, it provides a high-purity nucleotide analogue for enzymatic assays.

  • Concentration: 100 mM aqueous solution.
  • High purity: 99.94%.
  • Intended for research use only.
  • Function: chain termination of DNA polymerase in sequencing.
  • Form: liquid, colorless to light yellow.
  • Molecular weight 532.11 g·mol⁻¹; formula C10H14N2Na3O13P3.
  • Available in small prepared aliquots such as 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.