Learn More
Medchemexpress LLC ddTTP (trisodium) solution | 128524-26-5 | MFCD00079155 | 99.9% | 532.11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137694B100MG
Encompass_Preferred
ddTTP trisodium solution (100mM) is a 2',3'-dideoxyribonucleoside 5'-triphosphate (ddNTP). It functions as a chain-elongating inhibitor of DNA polymerase, making it a critical reagent for DNA sequencing applications. This product is intended for research use only.
  • Functions as a chain-elongating inhibitor of DNA polymerase
  • Utilized in DNA sequencing applications
  • Suitable for biochemical assays
  • Supports gene sequencing and synthesis
Catalog No. 50-003-52172