Learn More
Enzo Life Sciences ddWater (endotoxin-free) (sterile) (1.5ml). CAS: 7732-18-5
SDP
Catalog No. 502009345
Encompass_Preferred
Sterile, double distilled endotoxin-free water for use with TLRGRADE™ or endotoxin-free grade reagents. Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal. Alternative name: ddH2O pyrogen-free. Long Term Storage: +4°C.
Catalog No. 50-200-9345 Supplier Enzo Life Sciences Supplier No. ALX505008LD15
Sterile, double distilled endotoxin-free water for use with TLRGRADE™ or endotoxin-free grade reagents. Product has been subjected to multiple rounds of LPS removal by adsorption with activated charcoal. Alternative name: ddH2O pyrogen-free. Long Term Storage: +4°C.