Learn More
AdipoGen ddWater Endotoxin-free (sterile)
SDP
Catalog No. 501176382
Encompass_Preferred
ddWater Endotoxin-free (sterile), AdipoGen Life Sciences. Chemical. MW: . Endotoxin free diluent buffer used for the dilution of TLRpure LPS, MPLA and Lipid A and solubilisation of endotoxin-free reagents (e.g. ODNs).
Catalog No. 50-117-6382 Supplier AdipoGen Supplier No. IAX900002L010
ddWater Endotoxin-free (sterile), AdipoGen Life Sciences. Chemical. MW: . Endotoxin free diluent buffer used for the dilution of TLRpure LPS, MPLA and Lipid A and solubilisation of endotoxin-free reagents (e.g. ODNs).