Learn More
Aviva Systems Biology Corp Ddx20 Antibody - C-terminal region (ARP37239_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37239_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ddx20. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human Ddx20. Alternate names: GEMIN3, MGC159174, dp103ApplicationIHC, WB. Species reactivity: Mouse, Rat
Catalog No. 50-284-604