Learn More
Aviva Systems Biology Corp DDX21 Antibody - C-terminal region (ARP36375_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36375_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DDX21. Alternate names: GUA, GURDB, RH-II/GU, RH-II/GuAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-53043