Learn More
Aviva Systems Biology Corp DDX21 Antibody - N-terminal region (ARP36374_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36374_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX21. Alternate names: GUA, GURDB, RH-II/GU, RH-II/GuAApplicationWB. Species reactivity: Horse, Human, Rat, Yeast
Catalog No. 50-014-56133