Learn More
Aviva Systems Biology Corp DDX27 Antibody - N-terminal region (ARP36448_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36448_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX27. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX27. Alternate names: DKFZp667N057, FLJ12917, FLJ20596, FLJ22238, HSPC259, MGC1018, MGC163147, PP3241, RHLP, Rrp3p, dJ686N3.1, DRS1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56159