Learn More
Aviva Systems Biology Corp DDX28 Antibody - N-terminal region (ARP36457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX28. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX28. Alternate names: FLJ11282, MDDX28ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-56162