Learn More
Medchemexpress LLC HY-121832 5mg Medchemexpress, DDX3-IN-1 CAS:1919828-83-3 Purity:>98%
SDP
Catalog No. 501932561
Encompass_Preferred
Medchemexpress, HY-121832 5mg DDX3-IN-1 CAS:1919828-83-3 DDX3-IN-1 (Compound 16f) is a DEAD-box polypeptide 3 (DDX3) inhibitor with CC 50 s of 50 and 36 μM for HIV and HCV, respectively. Antiviral activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2561 Supplier Medchemexpress LLC Supplier No. HY1218325MG
Medchemexpress, HY-121832 5mg DDX3-IN-1 CAS:1919828-83-3 DDX3-IN-1 (Compound 16f) is a DEAD-box polypeptide 3 (DDX3) inhibitor with CC 50 s of 50 and 36 μM for HIV and HCV, respectively. Antiviral activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.