Learn More
Aviva Systems Biology Corp DDX39 Antibody - N-terminal region (ARP36384_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36384_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX39. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX39. Alternate names: BAT1, DDXL, BAT1L, DDX39, URH49ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53045