Learn More
Aviva Systems Biology Corp DDX3X Antibody - N-terminal region (ARP36342_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36342_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX3X. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX3X. Alternate names: DBX, DDX14, DDX3, HLP2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56119