Learn More
Eurofins Discoverx DDX3Y, active
Catalog No. 5000455604
Encompass
10 µg of active DDX3Y. N-terminal MBP-tagged, C-terminal 6His and Avi-tagged recombinant, full length, human DDX3Y.
Catalog No. 50-004-55604 Supplier Eurofins Discoverx Supplier No. 18023K
10 µg of active DDX3Y. N-terminal MBP-tagged, C-terminal 6His and Avi-tagged recombinant, full length, human DDX3Y.