Learn More
Eurofins Discoverx DDX3Y, active
Catalog No. 5000455605
Encompass
10 µg of active DDX3Y. N-terminal MBP-tagged, C-terminal 6His and Avi-tagged recombinant, full length, human DDX3Y.
Catalog No. 50-004-55605 Supplier Eurofins Discoverx Supplier No. 18023M
10 µg of active DDX3Y. N-terminal MBP-tagged, C-terminal 6His and Avi-tagged recombinant, full length, human DDX3Y.