Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci DDX41 Peptide
SDP

Catalog No. 16597223
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-597-223 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-597-223 Supplier ProSci Supplier No. 7349P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

DDX41 peptide is used for blocking the activity of DDX41 antibody (catalog number 7349). It usually blocks the antibody activity completely in Western blot by incubating the peptide with equal volume of antibody for 30 min at 37°C.

Specifications

Gene ID (Entrez) 51428
Species Human
Accession Number NP_057306
Common Name DEAD (Asp-Glu-Ala-Asp) box polypeptide 41
Concentration 200 μg/mL
Content And Storage Store DDX41 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol DDX41
Quantity 0.05 mg
Sequence 17 amino acid peptide near the amino terminus of human DD x 41.
Synonym ABS, ABS, Probable ATP-dependent RNA helicase DDX41, DEAD box protein 41
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.