Learn More
Aviva Systems Biology Corp DDX42 Antibody - N-terminal region (ARP36403_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36403_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX42. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX42. Alternate names: FLJ43179, RHELP, RNAHP, SF3b125ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56139