Learn More
Eurofins Discoverx DDX43, active
Catalog No. 5000455687
Encompass
10 µg of active DDX43. N-terminal 6His and Avi-tagged recombinant, full length, human DDX43.
Catalog No. 50-004-55687 Supplier Eurofins Discoverx Supplier No. 18015
10 µg of active DDX43. N-terminal 6His and Avi-tagged recombinant, full length, human DDX43.