Learn More
Aviva Systems Biology Corp DDX46 Antibody - C-terminal region (ARP37717_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37717_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DDX46. Alternate names: Prp5, PRPF5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53090