Learn More
Aviva Systems Biology Corp DDX46 Antibody - middle region (ARP36424_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36424_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX46. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DDX46. Alternate names: FLJ25329, KIAA0801, MGC9936, PRPF5, Prp5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56149