Learn More
Aviva Systems Biology Corp DDX47 Antibody - C-terminal region (ARP41210_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41210_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX47. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DDX47. Alternate names: RRP3, E4-DBP, HQ0256, MSTP162ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-57762