Learn More
Aviva Systems Biology Corp DDX47 Antibody - N-terminal region (ARP36439_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36439_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX47. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX47. Alternate names: DKFZp564O176, E4-DBP, FLJ30012, HQ0256, MSTP162, RRP3ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56154