Learn More
Eurofins Discoverx DDX49, active
Catalog No. 5000456030
Encompass
10 µg of active DDX49. N-terminal 6His and Avi-tagged recombinant, full length, human DDX49.
Catalog No. 50-004-56030 Supplier Eurofins Discoverx Supplier No. 18016M
10 µg of active DDX49. N-terminal 6His and Avi-tagged recombinant, full length, human DDX49.