Learn More
Aviva Systems Biology Corp DDX49 Antibody - N-terminal region (ARP36465_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36465_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX49. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX49. Alternate names: R27090_2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast, Zebrafish
Catalog No. 50-014-56164