Learn More
Aviva Systems Biology Corp DDX5 Antibody - N-terminal region (ARP36366_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36366_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX5. Alternate names: p68, HLR1, G17P1, HUMP68ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56129