Learn More
Aviva Systems Biology Corp DDX50 Antibody - N-terminal region (ARP36497_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36497_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX50. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX50. Alternate names: GU2, GUB, RH-II/GuBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56174