Learn More
Aviva Systems Biology Corp DDX51 Antibody - middle region (ARP36537_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36537_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX51. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human DDX51. Alternate names: DKFZp686N2081, MGC42193ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-499