Learn More
Aviva Systems Biology Corp DDX52 Antibody - N-terminal region (ARP36396_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36396_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX52. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX52. Alternate names: ROK1, HUSSY19ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56138