Learn More
Aviva Systems Biology Corp DDX54 Antibody - C-terminal region (ARP36498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX54. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DDX54. Alternate names: DP97, MGC2835ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56175