Learn More
Aviva Systems Biology Corp Ddx54 Antibody - C-terminal region (ARP37386_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37386_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ddx54. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2410015A15Rik, AI414901, DP97ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-622