Learn More
Aviva Systems Biology Corp DDX58 Antibody - middle region (ARP36416_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36416_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX58. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DDX58. Alternate names: DKFZp434J1111, DKFZp686N19181, FLJ13599, RIG-IApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-56146