Learn More
Aviva Systems Biology Corp DDX6 Antibody - N-terminal region (ARP36367_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36367_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DDX6. Alternate names: FLJ36338, HLR2, P54, RCKApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-492