Learn More
Chemglass Life Sciences Decapper, Portable, 11mm, for GC Chem Snap Caps
SDP
Catalog No. 501532666
Encompass_Preferred
GC Chem Snap Caps™ were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.
Catalog No. 50-153-2666 Supplier Chemglass Life Sciences Supplier No. CV57150011
GC Chem Snap Caps™ were developed to handle the volatile samples, providing a liquid airtight seal with the vial. Simply snap on the cap and an evaporation-proof seal of a crimped cap is achieved.