Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermco™ DE-Graph Windows Software

Catalog No. 04500319
Change view
Click to view available options
Dimensions (L x W x H):
125 mm x 125 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
04-500-319 125 mm x 125 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-500-319 Supplier Thermco™ Supplier No. ACCD0081PD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Accessory for P795

Thermco Products™ DE-Graph Windows Software is for use with P795 for online documentation

Specifications

Dimensions (L x W x H) 125 mm x 125 mm
For Use With (Application) Windows™
Weight (Metric) 25 g
Length (English) Yes
Description Windows Software DE-Graph: English, German and French. Online-documentation during measuring via PC. Simply memory download (DBF file format allows an easy data export to Excel). Graphic & table visualization of data.
For Use With (Equipment) ACCD795P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.