Learn More
Bioworld De Greef & Jacobs Medium, 50 L
Catalog No. 50254581
Encompass_Preferred
De Greef & Jacobs MediumContains Macronutrients and Micronutrients Sucrose, Cytokines, Vitamins, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L or 50L of medium.Plant tissue culture tested
Catalog No. 50-254-581 Supplier Bioworld Supplier No. 306300154
De Greef & Jacobs MediumContains Macronutrients and Micronutrients Sucrose, Cytokines, Vitamins, and Auxins should be added as neededProvided as 1 unit to make 10L, 25L or 50L of medium.Plant tissue culture tested