Learn More
Bioworld De Greef & Jacobs Medium w/ Vitamins, 50 L
Catalog No. 50254585
Encompass_Preferred
De Greef & Jacobs MediumContains Macronutrients, Micronutrients, and VitaminsWorking concentration; 3.91g/LProvided as 1 unit to make 10L (30630016-1), 25L or 50L of medium. Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-585 Supplier Bioworld Supplier No. 306300164
De Greef & Jacobs MediumContains Macronutrients, Micronutrients, and VitaminsWorking concentration; 3.91g/LProvided as 1 unit to make 10L (30630016-1), 25L or 50L of medium. Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested