Learn More
Bioworld De Greef and Jacobs Vitamin Mix, 100 mL
Catalog No. 50254587
Encompass_Preferred
De Greef & Jacobs Vitamin MixPackage contains 11.20 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/L Nicotinic Acid: 1.00 mg/L Pyridoxine HCl: 1.00 mg/L Thiamine HCl: 10.00 mg/L
Catalog No. 50-254-587 Supplier Bioworld Supplier No. 306300181
De Greef & Jacobs Vitamin MixPackage contains 11.20 g vitamins to prepare 100 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/L Nicotinic Acid: 1.00 mg/L Pyridoxine HCl: 1.00 mg/L Thiamine HCl: 10.00 mg/L