Learn More
Bioworld De Greef and Jacobs Vitamin Solution (Ampule), 100 mL
Catalog No. 50254586
Encompass_Preferred
De Greef & Jacobs Vitamin SolutionPackage includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/L Nicotinic Acid: 1.00 mg/L Pyridoxine HCl: 1.00 mg/L Thiamine HCl: 10.00 mg/L
Catalog No. 50-254-586 Supplier Bioworld Supplier No. 306300171
De Greef & Jacobs Vitamin SolutionPackage includes 10 prepared, filtered, and sterilized ampules of 10 mL of 1000X vitamin stock solution Use 1 mL per liter of mediumContents: myo-Inositol: 100.00 mg/L Nicotinic Acid: 1.00 mg/L Pyridoxine HCl: 1.00 mg/L Thiamine HCl: 10.00 mg/L