Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC De-N-sulfated heparin sodium salt | 61932-66-9 | MFCD00130910 | 95.4% | N/A | N/A | 5 MG
SDP

Catalog No. 5000283837
Encompass_Preferred

De-N-sulfated heparin sodium salt is a chemically modified heparin derivative used as a biochemical reagent in life-science research. Supplied as a white to off-white solid for research use only, it is suitable for biochemical assays and studies requiring N-desulfated heparin.

  • Purity: 95.4%.
  • CAS number: 61932-66-9.
  • Appearance: white to off-white solid.
  • Storage: powder stable at -20°C up to 3 years.
  • In solvent: store at -80°C up to 6 months or -20°C up to 1 month.
  • Available in multiple research-size vials.

Catalog No. 50-002-83837 Supplier Medchemexpress LLC Supplier No. HYW1456585MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

De-N-sulfated heparin sodium salt is a chemically modified heparin derivative used as a biochemical reagent in life-science research. Supplied as a white to off-white solid for research use only, it is suitable for biochemical assays and studies requiring N-desulfated heparin.

  • Purity: 95.4%.
  • CAS number: 61932-66-9.
  • Appearance: white to off-white solid.
  • Storage: powder stable at -20°C up to 3 years.
  • In solvent: store at -80°C up to 6 months or -20°C up to 1 month.
  • Available in multiple research-size vials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.