Learn More
Cayman Chemical DEA Buffer Concentrate (10X), 2.5 mL
SDP
Catalog No. 502942200
Encompass_Preferred
A buffer for development of alkaline phosphatase-based EIA kits.
Catalog No. 50-294-2200 Supplier Cayman Chemical Supplier No. 4000822.5 ML
A buffer for development of alkaline phosphatase-based EIA kits.