Learn More
Seracare Life Sciences Inc DEA BUFFER SOLUTION
Catalog No. 5067431
Encompass_Preferred
5M Diethanolamine (DEA) is used in conjunction with pNPP tablets to detect alkaline-phosphatase conjugates.
This item is not returnable. View return policy
Catalog No. 50-674-31 Supplier Seracare Life Sciences Inc Supplier No. 51200058
5M Diethanolamine (DEA) is used in conjunction with pNPP tablets to detect alkaline-phosphatase conjugates.