Learn More
Bioworld DEA Buffer (10X), 100 mL
Catalog No. 50255871
Encompass_Preferred
Diethanolamine Buffer Concentrate Product contains 50mL of 10X liquid concentrate and makes 500mL of 1X solution when diluted with distilled water.DEA, frequently used as a diluent for PNPP in ELISA assays Technical Information:Stability >1 year at 4°CContains diethanolamine and MgCl250 mLAppearance: LiquidBoiling Point : 217 °C (lit.) at 150 mmHgDensity : 1.10 g/mL at 25 °C (lit.)Melting Point: 28 °C (lit.)PH Value: 9.5RTECS Number: KL2975000
Catalog No. 50-255-871 Supplier Bioworld Supplier No. 404200002
Diethanolamine Buffer Concentrate Product contains 50mL of 10X liquid concentrate and makes 500mL of 1X solution when diluted with distilled water.DEA, frequently used as a diluent for PNPP in ELISA assays Technical Information:Stability >1 year at 4°CContains diethanolamine and MgCl250 mLAppearance: LiquidBoiling Point : 217 °C (lit.) at 150 mmHgDensity : 1.10 g/mL at 25 °C (lit.)Melting Point: 28 °C (lit.)PH Value: 9.5RTECS Number: KL2975000