Learn More
Cayman Chemical DEA NONOate, Purity: ≥98%, CAS: 372965-00-9, 100 mg
SDP
Catalog No. 502895016
Encompass_Preferred
A NO donor with a half-life of 2 minutes at 37°C, pH 7.4; liberates 1.5 moles of NO per mole of parent compound
Catalog No. 50-289-5016 Supplier Cayman Chemical Supplier No. 82100100 MG
A NO donor with a half-life of 2 minutes at 37°C, pH 7.4; liberates 1.5 moles of NO per mole of parent compound