Learn More
Cayman Chemical DEA Nonate 10mg
SDP
Catalog No. 502896320
Encompass_Preferred
A NO donor with a half-life of 2 minutes at 37°C, pH 7.4; liberates 1.5 moles of NO per mole of parent compound
Catalog No. 50-289-6320 Supplier Cayman Chemical Supplier No. 8210010 MG
A NO donor with a half-life of 2 minutes at 37°C, pH 7.4; liberates 1.5 moles of NO per mole of parent compound