Learn More
Cayman Chemical DEA NONOate, 372965-00-9, 100 mg
SDP
Supplier:  Cayman Chemical 82100100
Encompass_Preferred
Molecular Formula C4H11N3O2 • C4H11N, Purity ≥98%, Formula Weight 206.3, Synonyms: Diethylammonium (Z)-1-(N,N-diethylamino)diazen-1-ium-1,2-diolate, Diethylamine NONOate, DEA/NO; A NO donor with a half-life of 2 minutes at 37C, pH 7.4; liberates 1.5 moles of NO per mole of parent compound.
Catalog No. NC0793825