Learn More
Enzo Life Sciences DEA NONOate (10mg). CAS: 372965-00-9
SDP
Supplier:  Enzo Life Sciences ALX430034M010
Encompass_Preferred
Nitric oxide (NO) donor. Alternative name: DEA/NO, 2-(N,N-Diethylamino)-diazenolate-2-oxide . diethylammonium salt. Purity: ≥97% (NMR). Solubility: Soluble in 100% ethanol (25mg/ml), DMSO (2mg/ml), dimethyl formamide (2mg/ml) or PBS, pH 7.2 (10mg/ml). Long Term Storage: -80°C.
Catalog No. 50-200-9163