Learn More
Waters Corp Waters Torus OBD Preparative Column, DEA, 30 x 250 mm, 5 μm
Catalog No. NC1829009 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-density diol column is designed for chromatographic separations, offering additional selectivity and good peak shapes for acidic compounds.
  • Column chemistry: DEA (Diethylamine)
  • Dimension: OBD 30 x 250 mm
  • Particle size: 5 μm
Catalog No. NC1829009 Supplier Waters Corp Supplier No. 186008627
Add to Cart
Add to Cart
Request a Quote
This high-density diol column is designed for chromatographic separations, offering additional selectivity and good peak shapes for acidic compounds.
  • Column chemistry: DEA (Diethylamine)
  • Dimension: OBD 30 x 250 mm
  • Particle size: 5 μm