Learn More
Medchemexpress LLC Deac | 50995-74-9 | 99.9% | 250 MG
SDP
Catalog No. 5000414473
Encompass_Preferred
7ACC1 is a specific blocker for monocarboxylate transporter 1 (MCT-1) and MCT-4. It attenuates renal cancer cell proliferation, migration, invasion, and down-regulates MCT1/MCT4 expression and extracellular lactate, making it promising for cancer research.
  • Specific blocker for monocarboxylate transporter 1 and MCT-4
  • Attenuates renal cancer cell proliferation
  • Reduces cell migration and invasion
  • Down-regulates MCT1/MCT4 expression levels
  • Decreases extracellular lactate
  • Considered promising for cancer research
Catalog No. 50-004-14473 Supplier Medchemexpress LLC Supplier No. 250MGHYD0067
7ACC1 is a specific blocker for monocarboxylate transporter 1 (MCT-1) and MCT-4. It attenuates renal cancer cell proliferation, migration, invasion, and down-regulates MCT1/MCT4 expression and extracellular lactate, making it promising for cancer research.
  • Specific blocker for monocarboxylate transporter 1 and MCT-4
  • Attenuates renal cancer cell proliferation
  • Reduces cell migration and invasion
  • Down-regulates MCT1/MCT4 expression levels
  • Decreases extracellular lactate
  • Considered promising for cancer research