Learn More
Medchemexpress LLC HY-K0030 1 mL × 10 Medchemexpress, Deacetylase Inhibitor Cocktail (100× in 70% DMSO) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYK00301 ML W 10
Encompass_Preferred
Medchemexpress, HY-K0030 1 mL × 10 Deacetylase Inhibitor Cocktail (100× in 70% DMSO) CAS: Deacetylase Inhibitor Cocktail is a synergistic combination of chemicals designed to preserve the acetylation state of proteins. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3296