Learn More
Medchemexpress LLC HY-N0594 5mg Medchemexpress, Deacetylasperulosidic Acid CAS:14259-55-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05945MG
Encompass_Preferred
Medchemexpress, HY-N0594 5mg Deacetylasperulosidic Acid CAS:14259-55-3 Deacetylasperulosidic acid (DAA) is a major phytochemical constituent of Morinda citrifolia fruit. Deacetylasperulosidic acidhas antioxidant activity by increasing superoxide dismutase activity. Deacetylasperulosidic acid has anticlastogenic activity, suppressing the induction of chromosome aberrations in hamster ovary cells and mice. Deacetylasperulosidic acid prevents 4-nitroquinoline 1-oxide (4NQO) induced DNA damage in vitro, suppresses IL-2 production along with the activation of natural killer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2804