Learn More
Cayman Chemical DeacetylravIdomycIn 2.5mg
SDP
Catalog No. 502940094
Encompass_Preferred
A microbial metabolite with light-dependent antibiotic and anticancer activities; active against Gram-positive bacteria including B. subtilis, S. aureus, S. epidermidis, and E. faecalis (MICs = 0.78-1.56 and <0.006-0.049 μg/ml in the absence and presence of fluorescent light, respectively); reduces colony formation by human colon cancer cells in a clonogenic assay in a light-dependent manner
Catalog No. 50-294-0094 Supplier Cayman Chemical Supplier No. 279912.5 MG
A microbial metabolite with light-dependent antibiotic and anticancer activities; active against Gram-positive bacteria including B. subtilis, S. aureus, S. epidermidis, and E. faecalis (MICs = 0.78-1.56 and <0.006-0.049 μg/ml in the absence and presence of fluorescent light, respectively); reduces colony formation by human colon cancer cells in a clonogenic assay in a light-dependent manner